missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethylamine complex, 98+%

Product Code. 10604682 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10604682 25 g Glass bottle 25g
10361174 5 g Glass bottle 5g
10429490 100 g Glass bottle 100g
11912071 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10604682 Supplier Thermo Scientific Acros Supplier No. 177310250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74-94-2 | C2H10BN | 58.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-94-2
Molecular Formula C2H10BN
Molecular Weight (g/mol) 58.92
MDL Number MFCD00051068
InChI Key FTDUHBOCJSQEKS-UHFFFAOYSA-N
Synonym Dimethylamineborane, DMAB
IUPAC Name dimethylamine borane
SMILES B.CNC

Specifications

Melting Point 33.0°C to 37.0°C
Color White to Yellow
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CH3)2NH·BH3
Quantity 25 g
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,thf,,diglyme,benzene,toluene,chlorobenzene
Formula Weight 58.92
Percent Purity 98+%
Physical Form Flakes or Granules
Chemical Name or Material Borane-dimethylamine complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Borane-dimethylamine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.