missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 11300258
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-87-0
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
IUPAC Name boron;methylsulfanylmethane
SMILES B.CSC

Specifications

Melting Point -38°C
Color Colorless to Yellow
Density 0.801 g/mL
Boiling Point ∼44°C (decomposition)
Flash Point −1°C (31°F)
Odor Unpleasant
Packaging ChemSeal™ bottles
Linear Formula (CH3)2S·BH3
Refractive Index 1.457
Quantity 5 mL
UN Number UN2924
Beilstein 3663489
Sensitivity Air and moisture sensitive
Solubility Information Miscible with ethyl ether,tetrahydrofurane,dichloromethane,benzene,xylene,hexane,diglyme,dimethyl ether,ethyl acetate,toluene,methylene chloride and other aprotic solvents.
Formula Weight 75.97
Physical Form Liquid
Chemical Name or Material Borane-dimethyl sulfide complex
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'