missing translation for 'onlineSavingsMsg'
Learn More

Borane-dicyclohexylphosphine Complex, 95%, Thermo Scientific™

Product Code. 10361163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10361163 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10361163 Supplier Thermo Scientific Acros Supplier No. 380720010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 108756-88-3
Melting Point 77.0°C to 81.0°C
Color White
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C12H26BP
MDL Number MFCD03412072
Quantity 1 g
Synonym dicyclohexylphosphine-borane, dicyclohexylphosphane borane
Solubility Information Solubility in water: decomposes
InChI Key TVRLVVNHOYLIPR-UHFFFAOYSA-N
Molecular Weight (g/mol) 212.12
Formula Weight 212.12
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Borane-dicyclohexylphosphine complex
Show More Show Less
compliance-icons
Product Identifier
  • Borane-dicyclohexylphosphine complex
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P402-Store in a dry place.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.