missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bora Recombinant Rabbit Monoclonal Antibody (K01_1N63)
GREENER_CHOICE

Product Code. 30829919
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829919 100 μL
30828200 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829919 Supplier Invitrogen™ Supplier No. MA563290
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MGDVKESKMQ ITPETPGRIP VLNPFESPSD YSNLHEQTLA SPSVFKSTKL PTPGKFRWSI DQLAVINPVE IDPEDIHRQA LYLSHSRIDK DVEDKRQKAI EEFFTKDVIV PSPWTDHEGK QLSQCHSSKC TNINSDSPVG KKLTIHSEKS DAACQTLLSL PVDFNLENIL GDYFRADEFA DQSPGNLSS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%, Pig - 92%, Cynomolgus monkey - 97%.

Bora (Protein aurora borealis) is a key activator of Aurora Related Protein Kinase A (ARK-1), which is a centrosome-associated serine/threonine kinase that regulates centrosome maturation, bipolar spindle assembly and chromosome segregation during mitosis. Bora is localized to the nucleus until mitosis is initiated, then translocates to the cytoplasm in a Cdc2 dependent manner. Activation of Cdc2 initiates the release of Bora into the cytoplasm where it can bind and activate ARK-1. PLK1 (polo-like kinase-1) interacts with Bora to control the accessibility of its activation loop for phosphorylation and activation by ARK-1. Bora and ARK-1 cooperatively activate PLK1 and control mitotic entry. Bora mutants result in multipolar spindles in mitosis identical to those observed when ARK-1 function is blocked. Thus, the ARK1-Bora-PLK1 regulatory circuit in mammalian cells elucidates a key mechanism in cell cycle regulation. At least three isoforms of Bora are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bora
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1N63
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene BORA
Gene Accession No. Q5M864, Q6PGQ7, Q8BS90
Gene Alias 6720463M24Rik; AI317232; aurora borealis; BORA; bora, aurora kinase A activator; C13orf34; HsBora; Protein aurora borealis; RGD1309522; RP11-342J4.2
Gene Symbols BORA
Host Species Rabbit
Immunogen Recombinant protein of human Bora (aa 1-189).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306102, 77744, 79866
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.