Learn More
Invitrogen™ Bora Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589480
Description
Immunogen sequence: KYSLGSITSP SPISSPTFSP IEFQIGETPL SEQRKFTVHS PDASSGTNSN GITNPCIRSP YIDGCSPIKN WSPMRLQMYS GGTQYRTSVI QIPFTLETQG EDEEDKENIP STDVSSPAMD AAGIHLRQFS NEASTHGTHL VVTAMSVTQN QSSASEKELA LLQDVEREKD NNTVDMVDPI EIADETTWIK EPVDNGSLPM TDFVSGIAFS IENSHMCMSP LAESSVIPCE SSNIQMDSGY NTQNCGSNIM DTVGAESYCK ESDAQTCEVE SKSQAFNMKQ DHTTQRCWMK TASPFQCSSP; Positive Samples: HT-29, HeLa.
Bora (Protein aurora borealis) is a key activator of Aurora Related Protein Kinase A (ARK-1), which is a centrosome-associated serine/threonine kinase that regulates centrosome maturation, bipolar spindle assembly and chromosome segregation during mitosis. Bora is localized to the nucleus until mitosis is initiated, then translocates to the cytoplasm in a Cdc2 dependent manner. Activation of Cdc2 initiates the release of Bora into the cytoplasm where it can bind and activate ARK-1. PLK1 (polo-like kinase-1) interacts with Bora to control the accessibility of its activation loop for phosphorylation and activation by ARK-1. Bora and ARK-1 cooperatively activate PLK1 and control mitotic entry. Bora mutants result in multipolar spindles in mitosis identical to those observed when ARK-1 function is blocked. Thus, the ARK1-Bora-PLK1 regulatory circuit in mammalian cells elucidates a key mechanism in cell cycle regulation. At least three isoforms of Bora are known to exist.
Specifications
| Bora | |
| Polyclonal | |
| Unconjugated | |
| BORA | |
| 6720463M24Rik; AI317232; aurora borealis; BORA; bora, aurora kinase A activator; C13orf34; HsBora; Protein aurora borealis; RGD1309522; RP11-342J4.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79866 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.97 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6PGQ7 | |
| BORA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 260-559 of human BORA (NP_079084.4). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.