missing translation for 'onlineSavingsMsg'
Learn More

Bora Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18407283 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407283 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407283 Supplier Novus Biologicals Supplier No. NBP177346AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Bora Polyclonal antibody specifically detects Bora in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bora
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias aurora borealis, BORA, chromosome 13 open reading frame 34, FLJ22624, hsBora, protein aurora borealis
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human Bora. The immunogen is located within amino acids 410 - 460 of Bora. Amino Acid Squence: DVEREKDNNTVDMVD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 79866
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.