missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561611
This item is not returnable.
View return policy
Description
Immunogen sequence: PLEWYDDFPH VGYDLDGRRI YKPLRTRDEL DQFLDKMDDP DYWRTVQDPM TGRDLRLTDE QVALVRRLQS GQFGDVGFNP YEP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
Component of the PeBoW complex, which is required for maturation of 28S and 5.8S ribosomal RNAs and formation of the 60S ribosome.
Specifications
| BOP1 | |
| Polyclonal | |
| Unconjugated | |
| BOP1 | |
| AU020183; AW146150; block of proliferation 1; block of proliferation 1 protein; block of proliferation 1 protein {ECO:0000255; Bop1; D18861; Erb1p; HAMAP-Rule:MF_03027}; Kiaa0124; mKIAA0124; ribosome biogenesis protein BOP1; ribosome biogenesis protein BOP1 {ECO:0000255 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23246 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14137 | |
| BOP1 | |
| Recombinant protein corresponding to Human BOP1. Recombinant protein control fragment (Product #RP-99278). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction