missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15607
This item is not returnable.
View return policy
Description
BOP1 Polyclonal specifically detects BOP1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| BOP1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| block of proliferation 1, KIAA0124Block of proliferation 1 protein, ribosome biogenesis protein BOP1 | |
| Rabbit | |
| 84 kDa | |
| 0.1 mL | |
| Epigenetics | |
| 23246 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q14137 | |
| BOP1 | |
| Recombinant protein encompassing a sequence within the center region of human BOP1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Expected to cross react based on sequence identity: Rat (87%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction