missing translation for 'onlineSavingsMsg'
Learn More

BoNT-B Light Chain Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30731894 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731894 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731894 Supplier Novus Biologicals Supplier No. AF5420JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

BoNT-B Light Chain Polyclonal antibody specifically detects BoNT-B Light Chain in C. botulinum samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BoNT-B Light Chain
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias DcD
Host Species Sheep
Immunogen E. coli-derived recombinant Clostridium BoNT-B Light Chain, Pro2-His428, Accession # P10844
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6030102
Target Species C. botulinum
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.