missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bone Morphogenetic Protein 2, Human

Product Code. 15408709
Change view
Click to view available options
Quantity:
10μg
2μg
Unit Size:
10µg
2µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408709 10μg 10µg
15418709 2μg 2µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408709 Supplier Thermo Scientific Chemicals Supplier No. J67332.!E
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For research use

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

Formulation Lyophilized powder
Quantity 10μg
Source Escherichia coli
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Bone Morphogenetic Protein 2, Human
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.