Learn More
Restek™ Bonded Reversed Phase Resprep SPE Cartridges
Hydrophobic (nonpolar) silica-based adsorbents; used to extract hydrophobic analytes from polar matrices.
Brand:  Restek™ 24050
Code : Z3
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13179942
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Manufactured using high density polypropylene
  • May be processed by any of these techniques: positive pressure, sidearm flask, centrifuge or vacuum manifold
Specifications
Specifications
Hydrophobic (non-Polar)
Reversed Bonded
C18 (high load)
500 mg/3 mL
HDPE
3 mL, 500 mg
Silica
Used to Extract Hydrophobic Analytes From Polar Matrices, Such as Water (e.g., Pesticides From Water)
SPE Cartridge
Yes
50/Pk.