missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene Bonded Carbon Filter
AirClean™ Systems bonded carbon filters provide for a predictable and reliable gas phase filtration solution.
£940.00 - £1200.00
Specifications
Filter Type | Carbon |
---|---|
For Use With (Equipment) | Airclean |
Product Type | Carbon Filter |
Material | Activated Carbon |
For Use With (Application) | Chemical extraction |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17745924
|
Syngene
SYN-ACF121 |
ACF121 |
£1115.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17755924
|
Syngene
SYN-ACF12118 |
ACF12118 |
£1005.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17795924
|
Syngene
SYN-ACF141 |
ACF141 |
£1125.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17705934
|
Syngene
SYN-ACF14118 |
ACF14118 |
£940.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17755934
|
Syngene
SYN-ACF1G1 |
ACF1G1 |
£1145.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17765934
|
Syngene
SYN-ACF1G118 |
ACF1G118 |
£995.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17785964
|
Syngene
SYN-ACF43F |
ACF43F |
£1190.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17775984
|
Syngene
SYN-ACFGFG |
ACFGFG |
£1200.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Bonded layered carbon filter for organics and inorganics
- Even distribution of impregnates, if applicable
- Multi-layering options for larger application compatibility
Specifications
Carbon | |
Carbon Filter | |
Chemical extraction |
Airclean | |
Activated Carbon | |
Silver |