Learn More
Invitrogen™ Bolt™ Bis-Tris Welcome Pack, 12%
Description
The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 20 minutes.
The Welcome Pack contains:
• Mini Gel Tank (A25977)
• Bolt Bis-Tris Plus gels (2 boxes, 20 gels)
• Bolt MES Running Buffer, 20X (B0002)
• Bolt LDS Sample Buffer, 4X (B0007)
• Bolt Sample Reducing Agent, 10X (B0009)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
About the Mini Gel Tank
The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 20 minutes. Run times may vary depending on buffer conditions and power supplies used.
About Bolt Bis-Tris Plus Gels
Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8, 10, 12, and 4-12% gradient and multiple well formats. Bolt Bis-Tris gels feature wedge-shaped wells, which can easily load up to two times more sample volume allowing for detection of proteins in very dilute samples or visualization of low-abundance proteins.
For transferring your proteins to a membrane, we recommend using for Bolt Transfer Buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Warranty
Up to 12 months
Specifications
Specifications
| For Use With (Equipment) | Mini Gel Tank |
| Gel Percentage | 12% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Bis-Tris |
| Separation Range | 3.5 to 80 kDa |
| Wells | 15-well |
| Product Line | Bolt WedgeWell |
| Quantity | 1 Welcome Pk. kit |
| Shipping Condition | Approved for shipment at room temperature and wet ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.