missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOLL Monoclonal Antibody (OTI4C2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526649
This item is not returnable.
View return policy
Description
BOLL Monoclonal Antibody for Western Blot, IHC (P)
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| BOLL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N9W6 | |
| BOLL | |
| Full length human recombinant protein of BOLL produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C2 | |
| Unconjugated | |
| BOLL | |
| 4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 66037 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction