missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Boldenone

Product Code. 17231172 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 17231172

Brand: Thermo Scientific™ 465755000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Boldenone
CAS 846-48-0
Quantity 500 mg
Melting Point 168°C to 170°C
Molecular Formula C19 H26 O2
Formula Weight 286.41
Physical Form Powder or Crystals
Color White to Light-yellow to Light-beige
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.