missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOLA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562746
This item is not returnable.
View return policy
Description
Immunogen sequence: GLPLHHRMFA TQTEGELRVT QILKEKFPRA TAIKVTDISG GCGAMYEIKI ESEEFKEKRT VQQHQMVNQA LKEEIKEMHG LRIFTSVP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
This gene encodes a protein that plays an essential role in the production of iron-sulfur (Fe-S) clusters for the normal maturation of lipoate-containing 2-oxoacid dehydrogenases, and for the assembly of the mitochondrial respiratory chain complexes. Mutation in this gene has been associated with multiple mitochondrial dysfunctions syndrome-2. Two alternatively spliced transcript variants encoding different isoforms with distinct subcellular localization have been reported for this gene.
Specifications
| BOLA3 | |
| Polyclonal | |
| Unconjugated | |
| BOLA3 | |
| bolA family member 3; bolA homolog 3; BOLA3; bolA-like protein 3; MMDS2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 388962 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53S33 | |
| BOLA3 | |
| Recombinant protein fragment of Human BOLA3 (Product #RP-99684) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction