missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92289-0.02ml
This item is not returnable.
View return policy
Description
BOLA3 Polyclonal antibody specifically detects BOLA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| BOLA3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| bolA homolog 3 (E. coli), bolA-like 3, bolA-like protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-68 of human BOLA3 (NP_997717.2). MAAWSPAAAAPLLRGIRGLPLHHRMFATQTEGELRVTQILKEKFPRATAIKVTDISGGCGAMYEIKIE | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 388962 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction