missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BOLA2 Polyclonal specifically detects BOLA2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | BOLA2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | BolA Homolog 2B (E. Coli), BOLA2, BOLA2A, BOLA2B, BolA-Like 2B, BolA-Like 2B (E. Coli), BolA-Like Protein 2, BolA-Like Protein 2 Member B, BolA-Like Protein 2B |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the n terminal region of mouse BOLA2 (NP_780312). Peptide sequence MELSADYLREKLRQDLEAEHVEVEDTTLNRCATSFRVLVVSAKFEGKPLL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?