missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Tube Holder

Product Code. 10047751
Change view
Click to view available options
Tubing Size:
O.D. 1.6mm (1/16 in. ) and 3.2mm (1/8 in.)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
10047751 O.D. 1.6mm (1/16 in. ) and 3.2mm (1/8 in.) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10047751 Supplier Bohlender™ Supplier No. F70606
  • ONLINE EXCLUSIVE PRICE
    £37.30
    £28.81 / Pack of 1
    Save £8.49
    22% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with thermoelectric flanging tools. BOLA Tube Holders allow safe fixing of tubing during the flanging procedure.

  • For tubing with O.D. 1.6mm (1/16 in. ) and 3.2mm (1/8 in.)
    • Allows safe fixing of the tubing during the flanging procedure
      • Designed for easy assembly and handling
        • Avoid injuries due to the hot flanging tips

Compliance

flange, tube, holder, accessories, flanging

Specifications

For Use With (Application) Safe fixing of the tubing during the flanging procedure
Tubing Size O.D. 1.6mm (1/16 in. ) and 3.2mm (1/8 in.)
Product Type Tube Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.