Learn More
Bohlender™ BOLA™ Spherical Ground Joint Sleeves
Click to view available options
Standard Taper Joint No.:
S 13
S 19
S 29
S 40
Unit Size:
Pack of 2
Pack of 3
Description
- With gripping ring and sealing rings on the outside
- High-vacuum tight sealing
- Low leak rate: <1 x 0.01 mbar x l x s-1
- Solid construction for continued use
Specifications
Specifications
| Material | PTFE |
| Product Type | Ground Joint Sleeves |
| Standard Taper Joint No. | S 19 |