missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Spherical Ground Joint to GL Tube Fittings for S 29

Product Code. 17197294
Click to view available options
Thread Size:
GL 25
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17197294

Brand: Bohlender™ D79036

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For transition from spherical ground joints to GL threads.

  • Fitting made of PTFE
  • For transition from spherical ground joints to GL threads
  • For connection of hard-walled tubing (e.g. PTFE, PFA, FEP) with BOLA™ Laboratory Screw Joints
  • Product is only exposed to PTFE

Specifications

For Use With (Application) Connection of Spherical Ground Joints to GL Threads
Fits Tubing ID (English) 0.57 in.
Fits Tubing ID (Metric) 14.5 mm
Material PTFE
Product Type Bore Ø: 14.5 mm
Standard Taper Joint No. S 29
Thread Size GL 25
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.