Learn More
Bohlender™ BOLA™ Spare Knurled Nut
Product Code. 30274473
Click to view available options
Description:
Taper 1:10
Taper 1:10/1:10/1:10/1:5
Taper 1:10/1:10/1:5
Type:
Thread M 14
Thread M 7
Thread M 8
Closure Size:
NS 10
NS 12.5, NS 14.5, 13.5
NS 21.5, NS 24.0, NS 29.2, 28.0
Unit Size:
Pack of 20
Pack of 50
Product Code. 30274473
Brand: Bohlender™ J119841
Only null left
Add to basket
Add to basket
Spare knurled nut made of polypropylene.
  • For BOLA™ Stopcocks
  • Made of polypropylene
Specifications
Type Thread M 8
Description Taper 1:10/1:10/1:5
Material Polypropylene
Closure Size NS 12.5, NS 14.5, 13.5
Color Red