Learn More
Bohlender™ BOLA™ Screw In Stopcocks, 2-way
Product Code. 17176954
Click to view available options
Material:
PTFE
Unit Size:
Each
Product Code. 17176954
Brand: Bohlender™ B15602
2-way Screw In Stopcocks with male threads and cylindrical plug type.
  • For connection to screw-in threads on BOLA™ Reactor Lids to interrupt the flow of connected tubes
  • Available as 2-way stopcock with straight bore and one GL-threaded connection or as 3-way stopcock with L-bore and two GL-threaded connections
  • Cylindrical stopcock plug with grip made of PP for good tightness
  • Stop valve with mark of flow direction
  • Universal chemical resistance
  • Product is only exposed to PTFE
Specifications
Type 2-way Stopcock
Material PTFE
Diameter (Metric) Outside 6 mm
Diameter (English) Outside 0.24 in.
Size GL 18, Screw-in Thread NPT 1/2