Learn More
Bohlender™ BOLA™ Screw In Fittings GL
Product Code. 17146944
Click to view available options
Standard Taper Joint:
NPT 1
NPT 1/2
NPT 1/4
NPT 3/4
Size:
Wrench Size 15 mm
Wrench Size 22 mm
Wrench Size 32 mm
Wrench Size 45 mm
Type:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Product Code. 17146944
Brand: Bohlender™ B15214
Screw In Fittings with two male GL threads.
  • For connection to BOLA™ Reactor Lids as GL necks
  • With hexagonal gripping surface in standard wrench size
Specifications
Type GL 14
Material PTFE
Standard Taper Joint NPT 1/4
Size Wrench Size 15 mm