Learn More
Bohlender™ Bola PTFE Glass Stirrer Bearings
Product Code. 10751591
Click to view available options
Standard Taper Joint:
29/32
Unit Size:
Each
Product Code. 10751591
Brand: Bohlender™ C42509
Only null left
Add to basket
Add to basket
Use these bearings with a variety of stirrers. Bohlender™ Bola PTFE Glass Stirrer Bearings are comprised of a combination of a borosilicate glass piece with ground joint, an interior PTFE shaft guide with integrated special gasket and a GL screw cap made of PPS.
  • Gasket can be exchanged after wearing
  • Suitable for vacuum
  • Bearing for stirrer shafts made of stainless steel, glass and for BOLA Stirrer Shafts
Specifications
For Use With (Application) Suitable for vacuum, perfect bearing for Stainless Steel, glass and BOLA Stirrer Shafts
Quantity 1 Each
Standard Taper Joint 29/32
Temperature (Metric) -200°C to +250°C
Type Glass Stirrer Bearings
Material PTFE