Learn More
Bohlender™ BOLA™ Joint Clamps
Joint Clamps with steel spring core for connection of standard ground joints.
Brand: Bohlender™ H942-45
Additional Details : Weight : 0.24200kg
Description
- PTFE encapsulated steel spring core
- Universal chemical resistance since product is only exposed to PTFE
Specifications
PTFE | |
Connection of Standard Ground Joints | |
NS 45/50 |