Learn More
Bohlender™ BOLA™ Flat Flange Lids
Click to view available options
Joint Type:
DN 100
DN 150
DN 60
Standard Taper Joint:
NS 14/23, GL 18
NS 29/32
NS 29/32, NS 19/26, GL 25
Unit Size:
Each
Description
- Suitable for reaction vessels with SCHOTT flat flange
- Center neck socket NS 29/32
- Lateral necks with ground joint sockets or GL-thread
- Bottom side with centring collar for non-slipping alignment on the vessel
- All lateral connectors are made to insert tubing, tubes and probes either aside the center or vertical (parallel to the stirrer shaft) in order to avoid collisions
Specifications
Specifications
| Type | Flat Flange Lid |
| Material | PTFE |
| For Use With (Equipment) | Reaction Vessels with SCHOTT Flat Flange |
| Standard Taper Joint | NS 29/32, NS 19/26, GL 25 |
| Joint Type | DN 100 |
| Size | Lateral Necks: 2 x NS 29/32 Angled Aside the Shaft, 1 x NS 19/26 and 3 x GL 25 Vertical |