missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Double Sealing Cones for Connection Bolts

Product Code. 17177354
Change view
Click to view available options
For Use With:
Tubing with O.D. 1.6 mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
17177354 Tubing with O.D. 1.6 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177354 Supplier Bohlender™ Supplier No. F 83710
  • ONLINE EXCLUSIVE PRICE
    £31.65
    £20.71 / Pack of 5
    Save £10.94
    34% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Double sealing cones for connection bolts.

  • Replacement double sided sealing cone made of Poly Ether Ether Ketone (PEEK)
  • Suitable for connection bolts Cat. No. F 836-10

Specifications

For Use With (Application) Connection Bolts Cat. No. F 836-10
For Use With Tubing with O.D. 1.6 mm
Material PEEK
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.