missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Distributor with Stirrer Bearing

Product Code. 17177124
Click to view available options
Thread Size:
Cntr Neck GL25, Lateral Necks GL14
Cntr Neck GL32, Lateral Necks GL14
Diameter (Metric):
10 mm
16 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17177124

Brand: Bohlender™ C43510

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Distributor with Stirrer Bearings made of PTFE with ground joint cone NS 45/40.

  • Completely made of PTFE
  • With ground joint NS 45/40 and three GL-threaded necks
  • Center neck serves as stirrer bearing
  • A special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap provide a good sealing of the stirrer shaft
  • Lateral necks can be used for connection of tubes and tubing by means of BOLA™ Laboratory Screw Joints
  • Integrated special nut for unlocking of stuck ground joint components
  • Universal chemical resistance
  • Medium is only exposed to PTFE

Specifications

Cone Size NS 45/40
Diameter (English) 0.4 in.
Diameter (Metric) 10 mm
For Use With (Equipment) For Tubing with 8 mm Diameter
Thread Size Cntr Neck GL25, Lateral Necks GL14
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.