Learn More
Bohlender™ BOLA™ Conductors for Magnetic Stirrer Heads
Click to view available options
Cone Size:
NS 19/26
NS 45/40
Standard Taper Joint:
NS 19/26
NS 45/40
Thread Size:
Left-handed Thread M 14.5 x 1.5
Left-handed Thread M 21 x 1.75
Unit Size:
Each
Description
- Borosilicate glass with ground joint and glued upper bearing
- Alternatively without ground joint for fusing on the reactor lid (upper bearing to be glued is included).
Specifications
Specifications
| Cone Size | NS 19/26 |
| Standard Taper Joint | NS 19/26 |
| Material | Borosilicate Glass |
| Thread Size | Left-handed Thread M 14.5 x 1.5 |