missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Clamping Nut

Product Code. 17864064
Click to view available options
Diameter (Metric) Shaft:
4 mm
Unit Size:
Pack of 3
This item is not returnable. View return policy

Product Code. 17864064

Brand: Bohlender™ C90101

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Clamping Nut for Globe Stirrer Coupling.

  • Suitable for BOLA™ Globe Stirrer Couplings
  • Cat.-No. C 398-04

Specifications

Diameter (English) Shaft 0.16 in.
Diameter (Metric) Shaft 4 mm
For Use With (Equipment) BOLA™ Globe Stirrer Couplings
Quantity 3 Pk.
Material POM
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.