missing translation for 'onlineSavingsMsg'
Learn More

BOK Rabbit anti-Human, Mouse, Rat, Clone: S03-1B6, Novus Biologicals™

Product Code. 18397013
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397013 100 μg 100µL
18368425 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18397013 Supplier Bio-Techne Supplier No. NBP315008100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BOK Monoclonal antibody specifically detects BOK in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BOK
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone S03-1B6
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias Bcl2-L-9, BCL2L9bcl-2-related ovarian killer protein, Bcl-2-like protein 9, BCL2-related ovarian killer, BOKL, hBOK, MGC4631
Host Species Rabbit
Immunogen A synthetic peptide of human BOK (Uniprot#: Q9UMX3)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 666
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.