Learn More
Invitrogen™ BOK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120442
Description
Positive test controls include: Mouse brain, Rat testis. The target is usually found in the following locations: Endoplasmic reticulum membrane, Golgi apparatus membrane, Single-pass membrane protein. Immunogen sequence: MEVLRRSSVF AAEIMDAFDR SPTDKELVAQ AKALGREYVH ARLLRAGLSW SAPERAAPVP GRLAEVCAVL LRLGDELEMI RPSVYRNVAR QLHISLQSEP.
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains all four BCL-2 like domains and is a pro-apoptotic BCL-2 protein identified in the ovary.
Specifications
| BOK | |
| Polyclonal | |
| Unconjugated | |
| Bok | |
| AI847676; apoptosis activator Mtd; AU021146; BCL2 family apoptosis regulator BOK; BCL2 related ovarian killer; BCL2L9; bcl2-L-9; Bcl-2-like protein 9; BCL2-related ovarian killer; bcl-2-related ovarian killer protein; BCL2-related ovarian killer protein; BOK; BOK, BCL2 family apoptosis regulator; Bok-BH3; BOKL; hBOK; matador; mtd; Protein matador | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29884, 51800, 666 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35425, Q792S6, Q9UMX3 | |
| Bok | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human BOK (NP_115904.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.