missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA126427
This item is not returnable.
View return policy
Description
PA1-26427 detects Bok from rat samples.
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains all four BCL-2 like domains and is a pro-apoptotic BCL-2 protein identified in the ovary.
Specifications
| BOK | |
| Polyclonal | |
| Unconjugated | |
| Bok | |
| AI847676; apoptosis activator Mtd; AU021146; BCL2 family apoptosis regulator BOK; BCL2 related ovarian killer; BCL2L9; bcl2-L-9; Bcl-2-like protein 9; BCL2-related ovarian killer; bcl-2-related ovarian killer protein; BCL2-related ovarian killer protein; BOK; BOK, BCL2 family apoptosis regulator; Bok-BH3; BOKL; hBOK; matador; mtd; Protein matador | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29884, 51800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% Thimerosal; pH 7.2 | |
| O35425, Q792S6 | |
| Bok | |
| Synthetic peptide surrounding amino acid 44 of rat Bok. | |
| 50 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction