missing translation for 'onlineSavingsMsg'
Learn More

BOK Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

£462.00

Specifications

Antigen BOK
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18743113
View Documents
Novus Biologicals
NBP2-41045MFV610
0.1 mL
£462.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BOK Polyclonal antibody specifically detects BOK in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

BOK
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
mFluor Violet 610 SE
Rabbit
Apoptosis
Bcl2-L-9, BCL2L9bcl-2-related ovarian killer protein, Bcl-2-like protein 9, BCL2-related ovarian killer, BOKL, hBOK, MGC4631
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BOK. The immunogen is located within the first 50 amino acids of BOK. Amino Acid Squence: DAFDRSPTDKELVAQA
Primary
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human
666
IgG
Peptide affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.