missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Boc-Val-Leu-Gly-Arg-pNA Acetate Salt

Product Code. 16369845 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16369845 25 mg 25mg
16379845 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16369845 Supplier MP Biomedicals™ Supplier No. 08613081

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Boc-Val-Leu-Gly-Arg-pNA is a chromogenic substrate for horseshoe crab clotting enzyme for endotoxins detection.

  • Boc-VLGR-pNA acetate salt (sequence Boc-Val-Leu-Gly-Arg-pNA), is a chromogenic substrate for horseshoe crab clotting enzyme
  • It has been widely used in detection assays for bacterial endotoxins

Specifications

Content And Storage -20°C
Format Powder
For Use With (Application) Endotoxin detection
Molecular Weight (g/mol) 663.78
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.