missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC Polyclonal Antibody

BOC Polyclonal Antibody
Brand: Invitrogen PA566345
This item is not returnable.
View return policy
Description
BOC Polyclonal Antibody for ICC/IF
Specifications
| BOC | |
| Polyclonal | |
| Unconjugated | |
| Boc | |
| 4732455C11; biregional Cdon binding protein; biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein; BOC; BOC cell adhesion associated, oncogene regulated; Boc homolog; Boc homolog (mouse); brother of CDO; brother of CDON; CDON2; cell adhesion associated, oncogene regulated 2; mFLJ00376; Protein BOC; UNQ604/PRO1190 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BWV1 | |
| Boc | |
| Recombinant Human BOC. Recombinant protein control fragment (Product #RP-107029). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction