missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOC Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547308
This item is not returnable.
View return policy
Description
In Western blots, approximately 25% cross-reactivity with recombinant human BOC is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CDON and BOC are cell surface receptors of the immunoglobulin repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
Specifications
| BOC | |
| Polyclonal | |
| Unconjugated | |
| Boc | |
| 4732455C11; biregional Cdon binding protein; biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein; BOC; BOC cell adhesion associated, oncogene regulated; Boc homolog; Boc homolog (mouse); brother of CDO; brother of CDON; CDON2; cell adhesion associated, oncogene regulated 2; mFLJ00376; Protein BOC; UNQ604/PRO1190 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 117606 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q6AZB0 | |
| Boc | |
| Mouse myeloma cell line NS0-derived recombinant mouse BOC Asp26-Lys508 (Gln ins) Val509-Asp846. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction