missing translation for 'onlineSavingsMsg'
Learn More

(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%, Thermo Scientific Chemicals

Catalog Number p-4229095
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
89524-98-1
C10H20NO7P
297.24
MFCD02178893
LJHAPRKTPAREGO-ZETCQYMHSA-N
methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
397380
COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC
This item is not returnable. View return policy
89524-98-1
C10H20NO7P
297.24
MFCD02178893
LJHAPRKTPAREGO-ZETCQYMHSA-N
methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
397380
COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 69.7°C to 71.6°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
IUPAC Name methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
Formula Weight 297.25
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material (±)-BOC-α-Phosphonoglycine trimethyl ester
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'