missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-O-Benzyl-L-tyrosine, 99%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| CAS | 2130-96-3 |
| Melting Point | 112°C to 115°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (HPLC) |
| Molecular Formula | C21H25NO5 |
| MDL Number | MFCD00065597 |
| Quantity | 5 g |
| Specific Rotation | 27.3 |
| Synonym | boc-tyr bzl-oh, boc-o-benzyl-l-tyrosine, n-boc-o-benzyl-l-tyrosine, n-tert-butoxycarbonyl-o-benzyl-l-tyrosine, s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-benzyl-l-tyrisine, o-benzyl-n-tert-butoxycarbonyl-l-tyrosine, 2s-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, o-benzyl-n-tert-butoxy carbonyl-l-tyrosine, tyrosine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl |
| Show More |
Safety and Handling
Product Identifier
- BOC-O-Benzyl-L-tyrosine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction