missing translation for 'onlineSavingsMsg'
Learn More

BOC-O-Benzyl-L-threonine hydroxysuccinimide ester, 98%, Thermo Scientific™

£23.79 - £99.51

Chemical Identifiers

CAS 32886-43-4
Molecular Formula C20H26N2O7
Molecular Weight (g/mol) 406.44
MDL Number MFCD00037905
InChI Key SNEHQGZSMYSVFT-UHFFFAOYNA-N
Synonym boc-thr bzl-osu, boc-threonine bzl-osu, boc-o-benzyl-l-threonine hydroxysuccinimide ester, n-tert-butoxycarbonyl-o-benzyl-l-threonine succinimidyl ester, n-alpha-t-butyloxycarbonyl-o-benzyl-l-threonine succinimidyl ester, 2s,3r-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino butanoate, 2,5-dioxoazolidinyl 2s,3r-2-tert-butoxy carbonylamino-3-phenylmethoxy bu tanoate, 2,5-dioxopyrrolidin-1-yl 2s,3r-3-benzyloxy-2-tert-butoxycarbonyl amino butanoate
PubChem CID 12879029
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoate
SMILES CC(OCC1=CC=CC=C1)C(NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11965511
View Documents Promotion Details
Thermo Scientific Acros
440900010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11945511
View Documents Promotion Details
Thermo Scientific Acros
440900050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

32886-43-4
406.44
SNEHQGZSMYSVFT-UHFFFAOYNA-N
12879029
CC(OCC1=CC=CC=C1)C(NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
C20H26N2O7
MFCD00037905
boc-thr bzl-osu, boc-threonine bzl-osu, boc-o-benzyl-l-threonine hydroxysuccinimide ester, n-tert-butoxycarbonyl-o-benzyl-l-threonine succinimidyl ester, n-alpha-t-butyloxycarbonyl-o-benzyl-l-threonine succinimidyl ester, 2s,3r-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino butanoate, 2,5-dioxoazolidinyl 2s,3r-2-tert-butoxy carbonylamino-3-phenylmethoxy bu tanoate, 2,5-dioxopyrrolidin-1-yl 2s,3r-3-benzyloxy-2-tert-butoxycarbonyl amino butanoate
(2,5-dioxopyrrolidin-1-yl) (2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoate

Specifications

32886-43-4
White
97.5% min. (HPLC)
C20H26N2O7
10.6
SNEHQGZSMYSVFT-UHFFFAOYNA-N
(2,5-dioxopyrrolidin-1-yl) (2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoate
12879029
98%
BOC-O-Benzyl-L-threonine hydroxysuccinimide ester
104°C to 107°C
Authentic
Glass Bottle
MFCD00037905
boc-thr bzl-osu, boc-threonine bzl-osu, boc-o-benzyl-l-threonine hydroxysuccinimide ester, n-tert-butoxycarbonyl-o-benzyl-l-threonine succinimidyl ester, n-alpha-t-butyloxycarbonyl-o-benzyl-l-threonine succinimidyl ester, 2s,3r-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino butanoate, 2,5-dioxoazolidinyl 2s,3r-2-tert-butoxy carbonylamino-3-phenylmethoxy bu tanoate, 2,5-dioxopyrrolidin-1-yl 2s,3r-3-benzyloxy-2-tert-butoxycarbonyl amino butanoate
CC(OCC1=CC=CC=C1)C(NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
406.44
406.44
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BOC-O-Benzyl-L-threonine hydroxysuccinimide ester

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only