missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boc-NH-PEG3-OH, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 2-[2-(2-t-Boc-aminoethoxy]ethoxy]ethanol |
| CAS | 139115-92-7 |
| Molecular Formula | C11 H23 N O5 |
| Quantity | 50 mg |
| Synonym | N-[2-[2-(2-Hydroxyethoxy)ethoxy]ethyl]carbamic Acid 1,1-Dimethylethyl Ester, 2-[2-(2-tert-Butyloxycarbonylaminoethoxy)ethoxy]ethanol, 2-[2-(2-t-Boc-aminoethoxy]ethoxy]ethanol |
| InChI Formula | InChI=1S/C11H23NO5/c1-11(2,3)17-10(14)12-4-6-15-8-9-16-7-5-13/h13H,4-9H2,1-3H3,(H,12,14) |
| SMILES | CC(C)(C)OC(=O)NCCOCCOCCO |
| IUPAC Name | tert-butyl N-[2-[2-(2-hydroxyethoxy)ethoxy]ethyl]carbamate |
| Molecular Weight (g/mol) | 249.304 |
| Formula Weight | 249.1576 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction