missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boc-N-methylglycine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Boc-N-methylglycine |
| CAS | 13734-36-6 |
| Molecular Formula | C8H15NO4 |
| Quantity | 25 g |
| Synonym | N-Carboxy-sarcosine N-tert-Butyl Ester (8CI), N-[(1,1-Dimethylethoxy)carbonyl]-N-methylglycine, (t-Butoxycarbonyl)sarcosine, (tert-Butoxycarbonyl)sarcosine, (tert-Butyloxycarbonyl)sarcosine, 2-((t-Butoxycarbonyl)(methyl)amino)acetic Acid, 2-[(tert-Butoxycarbonyl)(methyl)amino]acetic Acid, Boc-sarcosine, N-(tert-Butoxycarbonyl)-N-methylglycine, N-(tert-Butoxycarbonyl)sarcosine, N-(tert- Butyloxycarbonyl)sarcosine, [(tert-Butoxycarbonyl)(methyl)amino]acetic Acid, [N-(tert-Butoxycarbonyl)-N-methylamino]acetic Acid |
| InChI Formula | InChI=1S/C8H15NO4/c1-8(2,3)13-7(12)9(4)5-6(10)11/h5H2,1-4H3,(H,10,11) |
| SMILES | CN(CC(=O)O)C(=O)OC(C)(C)C |
| IUPAC Name | 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid |
| Molecular Weight (g/mol) | 189.21 |
| Formula Weight | 189.1001 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction