missing translation for 'onlineSavingsMsg'
Learn More

BOC-α-Methylalanine, 98%, Thermo Scientific™

Product Code. 10574331 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10574331 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10574331 Supplier Thermo Scientific Acros Supplier No. 338790010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 30992-29-1
Assay Percent Range 98%
Molecular Formula C9H17NO4
Linear Formula (CH3)3COCONHC(CH3)2COOH
MDL Number MFCD00042973
Synonym boc-aib-oh, boc-alpha-methylalanine, 2-boc-amino isobutyric acid, 2-tert-butoxycarbonyl amino-2-methylpropanoic acid, boc-2-aminoisobutyric acid, n-boc-2-aminoisobutyric acid, n-boc-alpha-methylalanine, n-boc-2-amino-2-methylpropanoic acid, 2-tert-butoxy carbonyl amino-2-methylpropanoic acid, 2-tert-butoxycarbonylamino isobutyric acid
InChI Key MFNXWZGIFWJHMI-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC(C)(C)C(=O)O
IUPAC Name 2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molecular Weight (g/mol) 203.238
PubChem CID 2733845
Formula Weight 203.24
Percent Purity 98%
Chemical Name or Material BOC-α-Methylalanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.