missing translation for 'onlineSavingsMsg'
Learn More

Boc-Lys(Boc)-OSu, TRC

Product Code. 30336023
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30336023 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30336023

Brand: Toronto Research Chemicals TRCB6570605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dioxopyrrolidin-1-yl (2S)-2,6-Bis(tert-butoxycarbonylamino)hexanoate
CAS 30189-36-7
Molecular Formula C20 H33 N3 O8
Quantity 5 g
Synonym L-Lysine, N2,N6-bis[(1,1-dimethylethoxy)carbonyl]-, 2,5-dioxo-1-pyrrolidinyl ester, Carbamic acid, [(1S)-1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(1,1-dimethylethyl) ester (9CI), Carbamic acid, [1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-1,5-pentanediyl]bis-, bis(1,1-dimethylethyl) ester, (S)-, Succinimide, N-[(N2,N6-dicarboxy-L-lysyl)oxy]-, di-tert-butyl ester (8CI), 42: PN: US20070066537 PAGE: 13 claimed protein, Bis(tert-butoxycarbonyl)lysine-N-hydroxysuccinimide, Nα,Nω-Di(tert-butoxycarbonyl)-L-lysine N-hydroxysuccinimide ester
InChI Formula InChI=1S/C20H33N3O8/c1-19(2,3)29-17(27)21-12-8-7-9-13(22-18(28)30-20(4,5)6)16(26)31-23-14(24)10-11-15(23)25/h13H,7-12H2,1-6H3,(H,21,27)(H,22,28)/t13-/m0/s1
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
Molecular Weight (g/mol) 443.4913
Formula Weight 443.2268
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.