missing translation for 'onlineSavingsMsg'
Learn More

Boc-Leu-Arg-Arg-AMC2 HCl, TRC

Product Code. 30337821
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337821 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337821 Supplier Toronto Research Chemicals Supplier No. TRCB01526610MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boc-Leu-Arg-Arg-AMC Dihydrochloride
CAS 109358-46-5
Molecular Formula C33 H52 N10 O7
Quantity 10 mg
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-L-leucyl-L-arginyl-N-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-L-argininamide Dihydrochloride
InChI Formula InChI=1S/C33H52N10O7/c1-18(2)15-24(43-32(48)50-33(4,5)6)29(47)42-23(10-8-14-39-31(36)37)28(46)41-22(9-7-13-38-30(34)35)27(45)40-20-11-12-21-19(3)16-26(44)49-25(21)17-20/h11-12,16-18,22-24H,7-10,13-15H2,1-6H3,(H,40,45)(H,41,46)(H,42,47)(H,43,48)(H4,34,35,38)(H4,36,37,39)/t22-,23-,24-/m0/s1
SMILES CC(C)C[C@H](NC(=O)OC(C)(C)C)C(=O)N[C@@H](CCCN=C(N)N)C(=O)N[C@@H](CCCN=C(N)N)C(=O)Nc1ccc2C(=CC(=O)Oc2c1)C
IUPAC Name tert-butyl N-[(2S)-1-[[(2S)-5-(diaminomethylideneamino)-1-[[(2S)-5-(diaminomethylideneamino)-1-[(4-methyl-2-oxochromen-7-yl)amino]-1-oxopentan-2-yl]amino]-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]carbamate
Molecular Weight (g/mol) 700.829
Formula Weight 700.402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.