missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Serine methyl ester, 95%, Thermo Scientific Chemicals

Product Code. 11985481
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2766-43-0
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
InChI Key SANNKFASHWONFD-LURJTMIESA-N
Synonym boc-ser-ome, boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, n-boc-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-l-ser-ome, methyl tert-butoxycarbonyl-l-serinate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-ser-ome; boc-l-serine methyl ester
PubChem CID 7019177
IUPAC Name methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

Color Colorless to Yellow
Density 1.1000g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.1
Specific Rotation -18
Formula Weight 219.24
Percent Purity 95%
Physical Form Oil or Liquid
Chemical Name or Material BOC-L-Serine methyl ester
Show More Show Less
Product Identifier
  • BOC-L-Serine methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'