missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Serine methyl ester, 95%, Thermo Scientific Chemicals

Product Code. 11677978 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11677978 10 g Glass bottle 10g
11985481 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11677978 Supplier Thermo Scientific Acros Supplier No. 437820100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2766-43-0
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
InChI Key SANNKFASHWONFD-LURJTMIESA-N
Synonym boc-ser-ome, boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, n-boc-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-l-ser-ome, methyl tert-butoxycarbonyl-l-serinate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-ser-ome; boc-l-serine methyl ester
PubChem CID 7019177
IUPAC Name methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

Color Colorless to Yellow
Density 1.1000g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 10 g
Specific Gravity 1.1
Specific Rotation -18
Formula Weight 219.24
Percent Purity 95%
Physical Form Oil or Liquid
Chemical Name or Material BOC-L-Serine methyl ester
Show More Show Less
Product Identifier
  • BOC-L-Serine methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.