missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-L-Proline, 99+%, Thermo Scientific Chemicals
£67.70 - £195.00
Chemical Identifiers
CAS | 15761-39-4 |
---|---|
Molecular Formula | C10H17NO4 |
Molecular Weight (g/mol) | 215.249 |
MDL Number | MFCD00037324 |
InChI Key | ZQEBQGAAWMOMAI-ZETCQYMHSA-N |
Synonym | boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro |
PubChem CID | 85083 |
IUPAC Name | (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
SMILES | CC(C)(C)OC(=O)N1CCCC1C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10378960
|
Thermo Scientific Acros
275650250 |
25 g | Glass Bottle |
£67.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10016153
|
Thermo Scientific Acros
275651000 |
100 g | Glass Bottle |
£195.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15761-39-4 | |
215.249 | |
ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
85083 | |
CC(C)(C)OC(=O)N1CCCC1C(=O)O |
C10H17NO4 | |
MFCD00037324 | |
boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Specifications
133.0°C to 136.0°C | |
White | |
C10H17NO4 | |
MFCD00037324 | |
boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
CC(C)(C)OC(=O)N1CCCC1C(=O)O | |
− 60.50 | |
85083 | |
≥99% | |
Crystalline Powder or Crystals |
15761-39-4 | |
99+% | |
Authentic | |
22, 35 | |
ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
215.249 | |
215.25 | |
0.5% max. (K.F.) | |
BOC-L-Proline |
RUO – Research Use Only