missing translation for 'onlineSavingsMsg'
Learn More

Boc-L-Nitroarginine, TRC

Product Code. 30348291
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348291 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348291 Supplier Toronto Research Chemicals Supplier No. TRCB3916155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boc-L-Nitroarginine
CAS 2188-18-3
Molecular Formula C11H21N5O6
Quantity 5 g
Synonym Nalpha-tert-Butoxycarbonyl-NG-nitro-L-arginine, Nalpha-tert-Butoxycarbonyl-NG-nitroarginine, N2-[(1,1-Dimethylethoxy)carbonyl]-N5-[imino(nitroamino)methyl]-L-ornithine
InChI Formula InChI=1S/C11H21N5O6/c1-11(2,3)22-10(19)14-7(8(17)18)5-4-6-13-9(12)15-16(20)21/h7H,4-6H2,1-3H3,(H,14,19)(H,17,18)(H3,12,13,15)/t7-/m0/s1
SMILES CC(C)(C)OC(=O)N[C@@H](CCCNC(=N)N[N+](=O)[O-])C(=O)O
IUPAC Name (2S)-5-[[amino(nitramido)methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
Molecular Weight (g/mol) 319.31
Formula Weight 319.1492
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.